siRNA Details
| Gene Symbol: Tab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025729.4 | NP_080005.2 | 4 | 1168 | ||
| GenBank mRNA | AK220185.1 | - | 4 | 1168 | ||
| BC139024.1 | - | 3 | 1025 | 8862046 | ||
| BC139025.1 | - | 3 | 1025 | 8862047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tab3| Gene Symbol: Tab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025729.4 | NP_080005.2 | 4 | 1168 | ||
| GenBank mRNA | AK220185.1 | - | 4 | 1168 | ||
| BC139024.1 | - | 3 | 1025 | 8862046 | ||
| BC139025.1 | - | 3 | 1025 | 8862047 | ||