siRNA Details
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025730.3 | NP_080006.3 | 20 | 2735 | ||
GenBank mRNA | AK052591.1 | - | 20 | 2736 | ||
AY792512.1 | - | 20 | 2678 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025730.3 | NP_080006.3 | 20 | 2735 | ||
GenBank mRNA | AK052591.1 | - | 20 | 2736 | ||
AY792512.1 | - | 20 | 2678 |