siRNA Details
| Gene Symbol: Lrrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025730.3 | NP_080006.3 | 22 | 2895 | ||
| GenBank mRNA | AY792512.1 | - | 22 | 2838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrk2| Gene Symbol: Lrrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025730.3 | NP_080006.3 | 22 | 2895 | ||
| GenBank mRNA | AY792512.1 | - | 22 | 2838 | ||