siRNA Details
| Gene Symbol: Lrrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025730.3 | NP_080006.3 | 6 | 688 | ||
| GenBank mRNA | AK052591.1 | - | 6 | 689 | ||
| AY792512.1 | - | 6 | 631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrk2| Gene Symbol: Lrrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025730.3 | NP_080006.3 | 6 | 688 | ||
| GenBank mRNA | AK052591.1 | - | 6 | 689 | ||
| AY792512.1 | - | 6 | 631 | |||