siRNA Details
| Gene Symbol: Emc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025736.2 | NP_080012.1 | 8, 9 | 644 | ||
| GenBank mRNA | AF332065.1 | - | 8, 9 | 572 | ||
| AF332066.1 | - | 8, 9 | 573 | |||
| AK014989.1 | - | 8, 9 | 645 | |||
| AK015968.1 | - | 8, 9 | 610 | |||
| AK172887.1 | - | 8, 9 | 589 | |||
| BC004716.1 | - | 8, 9 | 602 | 3582196 | ||

Emc2
Genomic Map