siRNA Details
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025736.2 | NP_080012.1 | 1, 2 | 100 | ||
GenBank mRNA | AF332065.1 | - | 1, 2 | 28 | ||
AF332066.1 | - | 1, 2 | 28 | |||
AK014989.1 | - | 1, 2 | 101 | |||
AK015968.1 | - | 1, 2 | 66 | |||
AK171442.1 | - | 1, 2 | 89 | |||
AK172887.1 | - | 1, 2 | 45 | |||
BC004716.1 | - | 1, 2 | 58 | 3582196 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
