siRNA Details
| Gene Symbol: Cypt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025738.2 | NP_080014.2 | 2 | 660 | ||
| GenBank mRNA | BC061079.1 | - | 2 | 644 | 6773161 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cypt1| Gene Symbol: Cypt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025738.2 | NP_080014.2 | 2 | 660 | ||
| GenBank mRNA | BC061079.1 | - | 2 | 644 | 6773161 | |