siRNA Details
| Gene Symbol: Cfap97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025747.3 | NP_080023.1 | 2 | 1344 | ||
| GenBank mRNA | AK016776.1 | - | 3 | 1362 | ||
| AK048125.1 | - | 3 | 1429 | |||
| AK129001.1 | - | 2 | 779 | |||
| AK129358.1 | - | 3 | 1402 | |||
| BC049135.1 | - | 3 | 1387 | 6831539 | ||
| BC058092.1 | - | 2 | 1346 | 6821844 | ||

Cfap97
Genomic Map