siRNA Details
| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025749.3 | NP_080025.3 | 2 | 1313 | ||
| GenBank mRNA | AK007266.1 | - | 1 | 365 | ||
| AK016749.1 | - | 2 | 1314 | |||
| AY350709.1 | - | 4 | 1495 | |||
| BC055099.1 | - | 2 | 1309 | 6704066 | ||

Zfp474
Genomic Map