siRNA Details
| Gene Symbol: 4933417A18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025750.3 | NP_080026.2 | 1 | 272 | ||
| GenBank mRNA | AK016778.1 | - | 1 | 237 | ||
| AK016838.1 | - | 1 | 273 | |||
| BC100450.1 | - | 1 | 262 | 30919421 | ||
| BC145747.1 | - | 1 | 227 | 40131050 | ||

4933417A18Rik
Genomic Map