siRNA Details
| Gene Symbol: Gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025757.4 | NP_080033.3 | 5 | 1033 | ||
| GenBank mRNA | AJ404329.1 | - | 5 | 521 | ||
| AK012042.1 | - | 5 | 861 | |||
| AK017120.1 | - | 1033 | ||||
| AK039435.1 | - | 5 | 917 | |||
| AK077759.1 | - | 5 | 600 | |||
| AK142753.1 | - | 5 | 835 | |||
| AK164644.1 | - | 5 | 900 | |||
| BC044901.1 | - | 5 | 735 | 3660276 | ||
| BC046821.1 | - | 5 | 847 | 6400140 | ||
| BC052418.1 | - | 5 | 875 | 6401069 | ||

Gid4
Genomic Map