siRNA Details
| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025758.4 | NP_080034.2 | 3 | 809 | ||
| GenBank mRNA | AF425641.1 | - | 3 | 809 | ||
| AK017090.1 | - | 3 | 818 | |||
| AK164225.1 | - | 256 | ||||
| BC049572.1 | - | 3 | 814 | 6773716 | ||

Asb17
Genomic Map