siRNA Details
| Gene Symbol: MARCKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002356.6 | NP_002347.5 | 2 | 1932 | ||
| GenBank mRNA | AK074526.1 | - | 2 | 916 | ||
| BC089040.1 | - | 2 | 1779 | 6378349 | ||
| D10522.1 | - | 2 | 1889 | |||
| M68956.1 | - | 2 | 1832 | |||

MARCKS
Genomic Map