siRNA Details
| Gene Symbol: Olfr701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028910.3 | NP_083186.2 | 5 | 667 | ||
| GenBank mRNA | AK015248.1 | - | 4 | 611 | ||
| AK016560.1 | - | 5 | 666 | |||
| AK017036.1 | - | 4 | 560 | |||
| BC110461.2 | - | 4 | 526 | 40039685 | ||

Olfr701
Genomic Map