siRNA Details
| Gene Symbol: MARCKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002356.6 | NP_002347.5 | 2 | 2076 | ||
| GenBank mRNA | AK074526.1 | - | 2 | 1060 | ||
| BC089040.1 | - | 2 | 1923 | 6378349 | ||
| D10522.1 | - | 2 | 2033 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCKS| Gene Symbol: MARCKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002356.6 | NP_002347.5 | 2 | 2076 | ||
| GenBank mRNA | AK074526.1 | - | 2 | 1060 | ||
| BC089040.1 | - | 2 | 1923 | 6378349 | ||
| D10522.1 | - | 2 | 2033 | |||