siRNA Details
| Gene Symbol: Rbm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025776.2 | NP_080052.1 | 7 | 635 | ||
| GenBank mRNA | AK018452.1 | - | 7 | 658 | ||
| AK045116.1 | - | 7 | 655 | |||
| AK150271.1 | - | 7 | 648 | |||
| AK151758.1 | - | 7 | 648 | |||
| AK152252.1 | - | 7 | 648 | |||
| AK165277.1 | - | 7 | 653 | |||
| AK166600.1 | - | 7 | 656 | |||
| AK167509.1 | - | 7 | 675 | |||
| AK168919.1 | - | 7 | 636 | |||
| BC042691.1 | - | 7 | 654 | |||
| BC080205.1 | - | 7 | 633 | 3709760 | ||

Rbm22
Genomic Map