siRNA Details
| Gene Symbol: Bcl2l14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025778.3 | NP_080054.1 | 4 | 845 | ||
| GenBank mRNA | AK008682.1 | - | 3 | 770 | ||
| AK016670.1 | - | 4 | 846 | |||
| AK016997.1 | - | 4 | 845 | |||
| AK018579.1 | - | 3 | 812 | |||
| AK035707.1 | - | 3 | 777 | |||
| AK160247.1 | - | 4 | 848 | |||

Bcl2l14
Genomic Map