siRNA Details
| Gene Symbol: Thap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025780.3 | NP_080056.1 | 2 | 364 | ||
| GenBank mRNA | AK009162.1 | - | 2 | 343 | ||
| AK018577.1 | - | 2 | 344 | |||
| AK081303.1 | - | 2 | 365 | |||
| BC027502.1 | - | 2 | 272 | 1515752 | ||

Thap2
Genomic Map