siRNA Details
| Gene Symbol: Tmem170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025781.2 | NP_080057.1 | 3 | 1288 | ||
| GenBank mRNA | AK018496.1 | - | 3 | 1289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem170| Gene Symbol: Tmem170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025781.2 | NP_080057.1 | 3 | 1288 | ||
| GenBank mRNA | AK018496.1 | - | 3 | 1289 | ||