siRNA Details
| Gene Symbol: Tmem170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025781.2 | NP_080057.1 | 1 | 158 | ||
| GenBank mRNA | AK007926.1 | - | 1 | 202 | ||
| AK018496.1 | - | 1 | 159 | |||
| BC139377.1 | - | 1 | 233 | 8862399 | ||
| BC145964.1 | - | 1 | 233 | 40131267 | ||

Tmem170
Genomic Map