siRNA Details
| Gene Symbol: Ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133795.1 | NP_598556.1 | 7 | 805 | ||
| GenBank mRNA | AK077715.1 | - | 7 | 818 | ||
| AK166431.1 | - | 7 | 812 | |||
| AK170032.1 | - | 7 | 816 | |||
| AK170230.1 | - | 7 | 815 | |||
| BC010236.1 | - | 7 | 805 | 3581904 | ||

Ttc1
Genomic Map