siRNA Details
| Gene Symbol: Ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133795.1 | NP_598556.1 | 7, 8 | 829 | ||
| GenBank mRNA | AK077715.1 | - | 7, 8 | 842 | ||
| AK166431.1 | - | 7, 8 | 836 | |||
| AK170032.1 | - | 7, 8 | 840 | |||
| AK170230.1 | - | 7, 8 | 839 | |||
| BC010236.1 | - | 7, 8 | 829 | 3581904 | ||

Ttc1
Genomic Map