siRNA Details
| Gene Symbol: 0610009O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024179.5 | NP_077141.2 | 12 | 1459 | ||
| GenBank mRNA | AK002437.1 | - | 12 | 1449 | ||
| AK009400.1 | - | 5 | 471 | |||
| AK141675.1 | - | 1422 | ||||
| AK172900.1 | - | 12 | 1384 | |||

0610009O20Rik
Genomic Map