siRNA Details
| Gene Symbol: Wdr45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025793.3 | NP_080069.2 | 7, 8 | 866 | ||
| GenBank mRNA | AK002352.1 | - | 7, 8 | 827 | ||
| AK013170.1 | - | 7, 8 | 816 | |||
| AK146101.1 | - | 7, 8 | 867 | |||
| AK151975.1 | - | 7, 8 | 828 | |||
| AK168603.1 | - | 7, 8 | 819 | |||
| AK170602.1 | - | 7, 8 | 813 | |||
| BC004595.1 | - | 7, 8 | 810 | 3584839 | ||

Wdr45b
Genomic Map