siRNA Details
| Gene Symbol: Etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025794.2 | NP_080070.2 | 13 | 1947 | ||
| GenBank mRNA | AK002483.1 | - | 13 | 1900 | ||
| AK075673.1 | - | 13 | 1898 | |||
| AK141684.1 | - | 13 | 1901 | |||
| AK152624.1 | - | 13 | 1916 | |||
| AK169686.1 | - | 13 | 1852 | |||
| BC012522.1 | - | 13 | 1896 | 2648103 | ||
| BC057670.1 | - | 13 | 1914 | 2810976 | ||

Etfdh
Genomic Map