siRNA Details
| Gene Symbol: Trim35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029979.3 | NP_084255.2 | 2 | 552 | ||
| GenBank mRNA | AB060155.1 | - | 2 | 496 | ||
| AF145374.1 | - | 2 | 536 | |||
| AK003000.1 | - | 2 | 511 | |||
| AK032635.1 | - | 2 | 525 | |||
| AK075877.1 | - | 2 | 523 | |||
| AK163749.1 | - | 2 | 488 | |||
| BC049105.1 | - | 2 | 502 | 6418046 | ||
| BC053494.1 | - | 2 | 417 | |||
| BC145783.1 | - | 2 | 546 | 40131086 | ||
| BC145809.1 | - | 2 | 546 | 40131112 | ||

Trim35
Genomic Map