siRNA Details
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198294.3 | NP_938036.2 | 9 | 1326 | ||
GenBank mRNA | AK030022.1 | - | 6 | 930 | ||
BC079914.1 | - | 8 | 1129 | 30668628 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198294.3 | NP_938036.2 | 9 | 1326 | ||
GenBank mRNA | AK030022.1 | - | 6 | 930 | ||
BC079914.1 | - | 8 | 1129 | 30668628 |