siRNA Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025811.3 | NP_080087.1 | 3 | 1003 | ||
| GenBank mRNA | AK002650.1 | - | 3 | 588 | ||
| AK004564.1 | - | 3 | 650 | |||
| AK033384.1 | - | 3 | 1004 | |||
| BC095956.1 | - | 3 | 947 | 5330002 | ||
| BC138190.1 | - | 3 | 840 | 8861209 | ||
| BC138191.1 | - | 3 | 840 | 8861210 | ||

Nhlrc2
Genomic Map