siRNA Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025813.3 | NP_080089.1 | 10, 11 | 973 | ||
| GenBank mRNA | AK004586.1 | - | 10, 11 | 974 | ||
| AK032688.1 | - | 10, 11 | 941 | |||
| AK075859.1 | - | 10, 11 | 1003 | |||
| AK135235.1 | - | 10, 11 | 974 | |||
| AK136108.1 | - | 10, 11 | 965 | |||
| AK152444.1 | - | 10, 11 | 995 | |||
| AK159554.1 | - | 10, 11 | 1001 | |||
| AK166645.1 | - | 4, 5 | 365 | |||
| AK208838.1 | - | 2, 3 | 90 | |||
| BC024891.1 | - | 10, 11 | 955 | 3992247 | ||

Mfsd1
Genomic Map