siRNA Details
| Gene Symbol: Zfp869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039965.1 | NP_001035054.1 | 5 | 1721 | ||
| NM_001039967.1 | NP_001035056.1 | 6 | 1829 | |||
| NM_181274.3 | NP_851791.1 | 5 | 1789 | |||
| GenBank mRNA | AK004570.1 | - | 5 | 1732 | ||
| AK030679.1 | - | 6 | 1827 | |||
| AK083392.1 | - | 6 | 1744 | |||
| AK088889.1 | - | 4 | 1632 | |||
| AK136878.1 | - | 5 | 1725 | |||
| BC063330.1 | - | 1786 | ||||

Zfp869
Genomic Map