siRNA Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025826.4 | NP_080102.1 | 8 | 1086 | ||
| GenBank mRNA | AK004889.1 | - | 8 | 987 | ||
| AK076535.1 | - | 8 | 987 | |||
| AK131628.1 | - | 8 | 1033 | |||
| AK152325.1 | - | 8 | 975 | |||
| AK163322.1 | - | 4 | 567 | |||
| BC011235.1 | - | 8 | 968 | |||
| BC035231.1 | - | 8 | 952 | |||
| BC044826.1 | - | 8 | 1019 | |||
| BC054428.1 | - | 8 | 1028 | 5039627 | ||

Acadsb
Genomic Map