siRNA Details
| Gene Symbol: Lonp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168591.1 | NP_001162063.1 | 7 | 813 | ||
| NM_025827.3 | NP_080103.1 | 13 | 2117 | |||
| GenBank mRNA | AK004843.1 | - | 13 | 2031 | ||
| AK050231.1 | - | 13 | 2026 | |||
| AK075719.1 | - | 7 | 814 | |||
| AK088348.1 | - | 12 | 2023 | |||
| AK151103.1 | - | 13 | 2063 | |||
| AK151406.1 | - | 11 | 1631 | |||
| AK152964.1 | - | 11 | 1632 | |||
| AK168628.1 | - | 11 | 1641 | |||
| AK168791.1 | - | 13 | 2063 | |||
| AK168956.1 | - | 13 | 2063 | |||
| BC049090.1 | - | 13 | 2053 | 6401542 | ||

Lonp2
Genomic Map