siRNA Details
| Gene Symbol: Lman2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025828.3 | NP_080104.2 | 5, 6 | 876 | ||
| GenBank mRNA | AK004952.1 | - | 5, 6 | 707 | ||
| AK014384.1 | - | 5, 6 | 698 | |||
| AK083442.1 | - | 5, 6 | 708 | |||
| AK129028.1 | - | 5, 6 | 703 | |||
| AK150383.1 | - | 5, 6 | 705 | |||
| AK166953.1 | - | 5, 6 | 713 | |||
| AK169401.1 | - | 5, 6 | 704 | |||
| BC049221.1 | - | 5, 6 | 700 | 2599070 | ||
| BC055327.1 | - | 5, 6 | 876 | 5702182 | ||

Lman2
Genomic Map