siRNA Details
| Gene Symbol: Lman2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025828.3 | NP_080104.2 | 6 | 919 | ||
| GenBank mRNA | AK004952.1 | - | 6 | 750 | ||
| AK014384.1 | - | 6 | 741 | |||
| AK083442.1 | - | 6 | 751 | |||
| AK129028.1 | - | 6 | 746 | |||
| AK150383.1 | - | 6 | 748 | |||
| AK166953.1 | - | 6 | 756 | |||
| AK169401.1 | - | 6 | 747 | |||
| BC049221.1 | - | 6 | 743 | 2599070 | ||
| BC055327.1 | - | 6 | 919 | 5702182 | ||

Lman2
Genomic Map