siRNA Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025834.3 | NP_080110.1 | 7 | 797 | ||
| GenBank mRNA | AK005011.2 | - | 7 | 741 | ||
| AK008819.1 | - | 7 | 798 | |||
| BC024104.1 | - | 7 | 926 | 5136392 | ||
| BC038045.1 | - | 7 | 762 | 4196827 | ||

Proz
Genomic Map