siRNA Details
| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF373710.1 | - | 3, 4 | 196 | ||
| AK003572.2 | - | 3, 4 | 226 | |||
| AK003699.1 | - | 3, 4 | 150 | |||
| AK085669.1 | - | 3, 4 | 275 | |||
| BC013535.1 | - | 3, 4 | 159 | 4239189 | ||
| BY083019.1 | - | 3, 4 | 208 | |||

Vps28
Genomic Map