siRNA Details
| Gene Symbol: Myeov2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163425.1 | NP_001156897.1 | 2 | 125 | ||
| GenBank mRNA | AK003315.1 | - | 2 | 106 | ||
| AK012198.1 | - | 2 | 98 | |||
| AK152318.1 | - | 2 | 92 | |||
| BC049759.1 | - | 2 | 125 | |||
| BC147334.1 | - | 2 | 111 | 9056392 | ||
| BC147335.1 | - | 2 | 111 | 9056395 | ||
| BC147659.1 | - | 2 | 111 | 9087448 | ||
| BC147669.1 | - | 2 | 111 | 9087458 | ||
| BY631936.1 | - | 2 | 120 | |||

Myeov2
Genomic Map