siRNA Details
| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025846.2 | NP_080122.2 | 5 | 794 | ||
| GenBank mRNA | AK003733.1 | - | 2 | 180 | ||
| AK011419.1 | - | 5 | 794 | |||
| AK078870.1 | - | 5 | 795 | |||
| AK172037.1 | - | 5 | 786 | |||
| AK172113.1 | - | 5 | 2066 | |||
| BC003871.1 | - | 5 | 745 | 3492159 | ||

Rras2
Genomic Map