siRNA Details
| Gene Symbol: Sdhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025848.3 | NP_080124.1 | 4 | 488 | ||
| GenBank mRNA | AK013503.1 | - | 4 | 453 | ||
| AK013962.2 | - | 4 | 456 | |||
| AK135970.1 | - | 4 | 448 | |||
| BC145731.1 | - | 4 | 448 | 40131034 | ||
| BC145733.1 | - | 4 | 448 | 40131036 | ||

Sdhd
Genomic Map