siRNA Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025850.2 | NP_080126.1 | 2 | 146 | ||
| GenBank mRNA | AK005694.1 | - | 2 | 156 | ||
| AK017071.1 | - | 2 | 170 | |||
| AY819025.1 | - | 2 | 176 | |||
| BC046278.1 | - | 2 | 146 | 6310911 | ||

Fank1
Genomic Map