siRNA Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025857.2 | NP_080133.1 | 4 | 431 | ||
| GenBank mRNA | AK017714.1 | - | 4 | 400 | ||
| AK159897.1 | - | 4 | 432 | |||
| AK182339.1 | - | 3 | 186 | |||
| AK186664.1 | - | 3 | 186 | |||
| AK192186.1 | - | 3 | 187 | |||
| AK209997.1 | - | 3 | 223 | |||
| AK210563.1 | - | 3 | 186 | |||
| AK214885.1 | - | 3 | 223 | |||
| AK215257.1 | - | 3 | 223 | |||
| BC027311.1 | - | 4 | 427 | 3964905 | ||

Aagab
Genomic Map