siRNA Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025862.2 | NP_080138.2 | 8 | 927 | ||
| GenBank mRNA | AK009379.1 | - | 8 | 906 | ||
| AK019502.1 | - | 8 | 924 | |||
| AK029897.1 | - | 8 | 891 | |||
| AK049908.1 | - | 4 | 1243 | |||
| AK051091.1 | - | 8 | 882 | |||
| AK077335.1 | - | 8 | 907 | |||
| AK161255.1 | - | 8 | 881 | |||
| BC026535.1 | - | 8 | 901 | |||
| BC037644.1 | - | 8 | 903 | 5053565 | ||

Acad8
Genomic Map