siRNA Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025862.2 | NP_080138.2 | 10, 11 | 1240 | ||
| GenBank mRNA | AK009379.1 | - | 10, 11 | 1219 | ||
| AK019502.1 | - | 10, 11 | 1237 | |||
| AK029897.1 | - | 10, 11 | 1204 | |||
| AK049908.1 | - | 6, 7 | 1559 | |||
| AK051091.1 | - | 10, 11 | 1195 | |||
| AK077335.1 | - | 10, 11 | 1220 | |||
| AK161255.1 | - | 10, 11 | 1195 | |||
| AK189475.1 | - | 1, 2 | 29 | |||
| BC026535.1 | - | 10, 11 | 1213 | |||
| BC037644.1 | - | 10, 11 | 1216 | 5053565 | ||

Acad8
Genomic Map