siRNA Details
| Gene Symbol: Tmem206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025864.3 | NP_080140.1 | 6, 7 | 866 | ||
| GenBank mRNA | AK009526.1 | - | 6, 7 | 867 | ||
| AK012107.1 | - | 6, 7 | 884 | |||
| AK031270.1 | - | 6, 7 | 900 | |||
| AK044482.1 | - | 6, 7 | 867 | |||
| AK049641.1 | - | 6, 7 | 903 | |||
| BC027151.1 | - | 6, 7 | 860 | 4948749 | ||

Tmem206
Genomic Map