siRNA Details
| Gene Symbol: Tmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290751.1 | NP_001277680.1 | 6 | 728 | ||
| NM_025868.4 | NP_080144.1 | 8 | 1028 | |||
| GenBank mRNA | AK009759.1 | - | 7 | 716 | ||
| AK075750.1 | - | 8 | 1030 | |||
| AK145245.1 | - | 7 | 737 | |||
| AK170587.1 | - | 7 | 725 | |||
| BC013544.1 | - | 7 | 710 | 4242418 | ||
| BC110994.1 | - | 7 | 711 | 5349078 | ||

Tmx2
Genomic Map