siRNA Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024185.4 | NP_077147.2 | 11, 12 | 1154 | ||
| GenBank mRNA | AK002900.1 | - | 11, 12 | 1105 | ||
| AK009891.1 | - | 11, 12 | 1133 | |||
| AK010050.1 | - | 11, 12 | 1120 | |||
| AK049869.1 | - | 10, 11 | 1078 | |||
| AK075792.1 | - | 9, 10 | 924 | |||
| AK143250.1 | - | 11, 12 | 1126 | |||
| BC002225.1 | - | 11, 12 | 1136 | |||
| BC027202.1 | - | 11, 12 | 1058 | 4217391 | ||

Fam188a
Genomic Map