siRNA Details
| Gene Symbol: Ssbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024186.5 | NP_077148.1 | 4 | 352 | ||
| NM_024272.4 | NP_077234.2 | 4 | 352 | |||
| GenBank mRNA | AK005150.1 | - | 4 | 287 | ||
| AK010220.1 | - | 4 | 294 | |||
| AK013177.1 | - | 4 | 278 | |||
| AK039140.1 | - | 924 | ||||
| AK051840.1 | - | 4 | 289 | |||
| AK133699.1 | - | 4 | 296 | |||
| AY037837.1 | - | 4 | 308 | |||
| AY167988.1 | - | 4 | 262 | |||
| BC002176.1 | - | 4 | 364 | 3486988 | ||

Ssbp2
Genomic Map