siRNA Details
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024186.5 | NP_077148.1 | 7, 8 | 581 | ||
NM_024272.4 | NP_077234.2 | 8, 9 | 671 | |||
GenBank mRNA | AK010220.1 | - | 7, 8 | 523 | ||
AK013177.1 | - | 8, 9 | 597 | |||
AK133699.1 | - | 7, 8 | 525 | |||
AY037837.1 | - | 8, 9 | 627 | |||
AY167988.1 | - | 8, 9 | 581 | |||
BC002176.1 | - | 7, 8 | 593 | 3486988 | ||
EF410127.1 | - | 4, 5 | 553 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760652, XM_008760651, XM_008760653, XM_008760650, XM_008760646, and others.
Panther Classification:
Gene Ontology Categories:
