siRNA Details
| Gene Symbol: Trappc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093759.1 | NP_001087228.1 | 4 | 584 | ||
| NM_001093760.1 | NP_001087229.1 | 4 | 584 | |||
| NM_025879.2 | NP_080155.3 | 4 | 584 | |||
| GenBank mRNA | AK010346.1 | - | 4 | 565 | ||
| AK013956.1 | - | 4 | 542 | |||
| AK084373.1 | - | 4 | 534 | |||
| AK149832.1 | - | 4 | 587 | |||
| AK163342.1 | - | 4 | 572 | |||
| AK167749.1 | - | 4 | 544 | |||
| BC021756.1 | - | 4 | 525 | 3584010 | ||

Trappc13
Genomic Map