siRNA Details
| Gene Symbol: Luc7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025881.3 | NP_080157.1 | 9 | 1007 | ||
| NM_028190.3 | NP_082466.2 | 9 | 1007 | |||
| GenBank mRNA | AK010539.1 | - | 9 | 959 | ||
| AK017655.1 | - | 9 | 967 | |||
| BC019194.1 | - | 10 | 1000 | |||
| BC034135.1 | - | 8 | 1173 | 5003664 | ||
| BC055875.1 | - | 9 | 1990 | |||
| CF951283.1 | - | 5 | 403 | |||

Luc7l
Genomic Map